Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144964.1 | NP_001138436.1 | 14 - 15 | 1490 | 113 |
NM_001144965.1 | NP_001138437.1 | 14 - 15 | 1442 | 113 | |
NM_001144966.2 | NP_001138438.1 | 13 - 14 | 1367 | 113 | |
NM_001144967.2 | NP_001138439.1 | 14 - 15 | 1536 | 113 | |
NM_001144968.1 | NP_001138440.1 | 14 - 15 | 1432 | 113 | |
NM_001144969.1 | NP_001138441.1 | 13 - 14 | 1372 | 113 | |
NM_001144970.2 | NP_001138442.1 | 12 - 13 | 1415 | 113 | |
NM_001144971.1 | NP_001138443.1 | 14 - 15 | 1460 | 113 | |
NM_015277.5 | NP_056092.2 | 13 - 14 | 1476 | 113 | |
XM_005266658.4 | XP_005266715.2 | 10 - 11 | 1413 | 113 | |
XM_005266660.4 | XP_005266717.2 | 10 - 11 | 1411 | 113 | |
XM_005266663.4 | XP_005266720.2 | 9 - 10 | 1351 | 113 | |
XM_006722421.3 | XP_006722484.2 | 9 - 10 | 1352 | 113 | |
XM_006722426.3 | XP_006722489.1 | 15 - 16 | 1630 | 113 | |
XM_006722428.3 | XP_006722491.1 | 14 - 15 | 1552 | 113 | |
XM_006722430.3 | XP_006722493.1 | 13 - 14 | 1460 | 113 | |
XM_011525887.2 | XP_011524189.1 | 15 - 16 | 1610 | 113 | |
XM_017025678.1 | XP_016881167.1 | 13 - 14 | 1491 | 113 | |
XM_017025679.1 | XP_016881168.1 | 13 - 14 | 1460 | 113 | |
GenBank mRNA | AB007899.1 | - | 12 - 13 | 1315 | 113 |
AB071179.1 | - | 13 - 14 | 1307 | 113 | |
AF210730.1 | - | 13 - 14 | 1267 | 113 | |
AF385931.1 | - | 14 - 15 | 1491 | 113 | |
AK294963.1 | - | 12 - 13 | 1364 | 113 | |
AK300444.1 | - | 13 - 14 | 1318 | 113 | |
AY112983.1 | - | 14 - 15 | 1442 | 113 | |
AY112984.2 | - | 14 - 15 | 1432 | 113 | |
AY112985.2 | - | 13 - 14 | 1376 | 113 | |
AY312514.1 | - | 14 - 15 | 1253 | 113 | |
BC000621.2 | - | 8 - 9 | 893 | 113 | |
BC019345.1 | - | 8 - 9 | 893 | 113 | |
DQ181796.1 | - | 14 - 15 | 1460 | 113 |