Assay Design
Gene Transcripts
- Gene Symbol
- NEDD4L
- Entrez Gene ID
-
23327
- Gene Name
- neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase
- Gene Aliases
-
NEDD4-2, NEDD4.2, RSP5, hNEDD4-2
- Chromosome Location
-
Chr.18: 58044362 - 58401540 on Build GRCh38
View Full Product Details