Assay Design



Gene Transcripts

Gene Symbol
NEDD4L
Entrez Gene ID
23327
Gene Name
neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase
Gene Aliases
NEDD4-2, NEDD4.2, RSP5, hNEDD4-2
Chromosome Location
Chr.18: 58044362 - 58401540 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144964.1 NP_001138436.1 9 - 10 914 88
NM_001144965.1 NP_001138437.1 9 - 10 866 88
NM_001144966.2 NP_001138438.1 8 - 9 791 88
NM_001144967.2 NP_001138439.1 9 - 10 960 88
NM_001144968.1 NP_001138440.1 9 - 10 856 88
NM_001144969.1 NP_001138441.1 9 - 10 856 88
NM_001144970.2 NP_001138442.1 8 - 9 899 88
NM_001144971.1 NP_001138443.1 10 - 11 944 88
NM_001243960.1 NP_001230889.1 9 - 10 960 88
NM_015277.5 NP_056092.2 9 - 10 960 88
XM_005266658.4 XP_005266715.2 5 - 6 837 88
XM_005266660.4 XP_005266717.2 5 - 6 835 88
XM_005266663.4 XP_005266720.2 5 - 6 835 88
XM_006722421.3 XP_006722484.2 5 - 6 836 88
XM_006722424.3 XP_006722487.2 5 - 6 839 88
XM_006722425.3 XP_006722488.2 5 - 6 837 88
XM_006722426.3 XP_006722489.1 10 - 11 1054 88
XM_006722428.3 XP_006722491.1 9 - 10 976 88
XM_006722430.3 XP_006722493.1 8 - 9 884 88
XM_011525887.2 XP_011524189.1 10 - 11 1034 88
XM_017025676.1 XP_016881165.1 5 - 6 838 88
XM_017025677.1 XP_016881166.1 5 - 6 836 88
XM_017025678.1 XP_016881167.1 9 - 10 975 88
XM_017025679.1 XP_016881168.1 8 - 9 884 88
XM_017025680.1 XP_016881169.1 8 - 9 884 88
XM_017025681.1 XP_016881170.1 8 - 9 884 88
GenBank mRNA AB007899.1 - 8 - 9 799 88
AB071179.1 - 9 - 10 791 88
AF210730.1 - 8 - 9 691 88
AF385931.1 - 9 - 10 915 88
AK294963.1 - 8 - 9 848 88
AK300444.1 - 8 - 9 742 88
AL137469.1 - 7 - 8 524 88
AY112983.1 - 9 - 10 866 88
AY112984.2 - 9 - 10 856 88
AY112985.2 - 9 - 10 860 88
AY312514.1 - 9 - 10 677 88
BC000621.2 - 4 - 5 377 88
BC019345.1 - 4 - 5 377 88
BC032597.1 - 9 - 10 987 88
DQ181796.1 - 10 - 11 944 88

View Full Product Details