Assay Design



Gene Transcripts

Gene Symbol
ARMC3
Entrez Gene ID
219681
Gene Name
armadillo repeat containing 3
Gene Aliases
CT81, KU-CT-1
Chromosome Location
Chr.10: 22928024 - 23038527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282745.1 NP_001269674.1 4 - 5 369 133
NM_001282746.1 NP_001269675.1 4 - 5 369 133
NM_001282747.1 NP_001269676.1 3 - 4 251 133
NM_173081.4 NP_775104.2 4 - 5 369 133
XM_005252380.2 XP_005252437.1 4 - 5 352 133
XM_005252381.2 XP_005252438.1 3 - 4 190 133
XM_011519350.2 XP_011517652.1 4 - 5 352 133
XM_017015831.1 XP_016871320.1 4 - 5 352 133
XM_017015832.1 XP_016871321.1 4 - 5 352 133
XM_017015833.1 XP_016871322.1 4 - 5 352 133
GenBank mRNA AK057389.1 - 4 - 5 312 133
AK090961.1 - 2 - 3 313 133
AK302104.1 - 3 - 4 198 133
AK302985.1 - 4 - 5 356 133
AY436643.1 - 4 - 5 330 133
BC039312.1 - 4 - 5 362 133
DB459427.1 - 4 - 5 369 133

View Full Product Details