Assay Design



Gene Transcripts

Gene Symbol
ARMC3
Entrez Gene ID
219681
Gene Name
armadillo repeat containing 3
Gene Aliases
CT81, KU-CT-1
Chromosome Location
Chr.10: 22928024 - 23038527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282745.1 NP_001269674.1 8 - 9 997 127
NM_001282746.1 NP_001269675.1 8 - 9 997 127
NM_001282747.1 NP_001269676.1 5 - 6 508 127
NM_173081.4 NP_775104.2 8 - 9 997 127
XM_005252380.2 XP_005252437.1 7 - 8 785 127
XM_005252381.2 XP_005252438.1 7 - 8 818 127
XM_011519350.2 XP_011517652.1 8 - 9 980 127
XM_017015831.1 XP_016871320.1 7 - 8 785 127
XM_017015832.1 XP_016871321.1 8 - 9 980 127
XM_017015833.1 XP_016871322.1 7 - 8 785 127
GenBank mRNA AK057389.1 - 8 - 9 940 127
AK302104.1 - 5 - 6 455 127
AK302985.1 - 8 - 9 984 127
AY436643.1 - 8 - 9 958 127
BC039312.1 - 8 - 9 990 127

View Full Product Details