Assay Design



Gene Transcripts

Gene Symbol
EPYC
Entrez Gene ID
1833
Gene Name
epiphycan
Gene Aliases
DSPG3, PGLB, Pg-Lb, SLRR3B
Chromosome Location
Chr.12: 90963679 - 91005026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004950.4 NP_004941.2 2 - 3 256 96
GenBank mRNA AK290642.1 - 2 - 3 213 96
BC030958.1 - 2 - 3 96
KM111190.1 - 2 - 3 247 96

View Full Product Details