Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018686.5 | NP_061156.1 | 4 - 5 | 823 | 129 |
NR_135117.1 | 4 - 5 | 823 | 129 | ||
GenBank mRNA | AF271388.1 | - | 4 - 5 | 711 | 129 |
AF397212.1 | - | 4 - 5 | 693 | 129 | |
AK022927.1 | - | 4 - 5 | 733 | 129 | |
AK222981.1 | - | 4 - 5 | 722 | 129 | |
AL832975.1 | - | 4 - 5 | 771 | 129 | |
BC016609.1 | - | 4 - 5 | 772 | 129 |