Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004561.3 | NP_004552.2 | 2 - 3 | 663 | 67 |
XM_005274018.3 | XP_005274075.1 | 2 - 3 | 481 | 67 | |
XM_011545067.2 | XP_011543369.1 | 2 - 3 | 479 | 67 | |
XM_011545068.2 | XP_011543370.1 | 2 - 3 | 1024 | 67 | |
XM_017017837.1 | XP_016873326.1 | 2 - 3 | 1317 | 67 | |
XM_017017838.1 | XP_016873327.1 | 2 - 3 | 356 | 67 | |
GenBank mRNA | AF016045.1 | - | 1 - 2 | 543 | 67 |
AK291711.1 | - | 2 - 3 | 598 | 67 | |
BC043146.1 | - | 2 - 3 | 254 | 67 | |
BC059408.1 | - | 2 - 3 | 639 | 67 | |
BX648725.1 | - | 2 - 3 | 543 | 67 |