Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100398.1 | NP_001093868.1 | 21 - 22 | 2155 | 142 |
NM_015085.4 | NP_055900.4 | 22 - 23 | 2200 | 142 | |
XM_005256542.2 | XP_005256599.1 | 21 - 22 | 2592 | 142 | |
XM_006721477.3 | XP_006721540.1 | 22 - 23 | 2141 | 142 | |
XM_011523738.1 | XP_011522040.1 | 23 - 24 | 2253 | 142 | |
XM_011523739.1 | XP_011522041.1 | 22 - 23 | 2208 | 142 | |
XM_011523740.2 | XP_011522042.1 | 22 - 23 | 2113 | 142 | |
XM_011523741.2 | XP_011522043.1 | 22 - 23 | 2289 | 142 | |
XM_011523742.2 | XP_011522044.1 | 19 - 20 | 1995 | 142 | |
XM_011523743.2 | XP_011522045.1 | 20 - 21 | 2073 | 142 | |
XM_011523744.2 | XP_011522046.1 | 15 - 16 | 1661 | 142 | |
XM_011523745.2 | XP_011522047.1 | 16 - 17 | 1748 | 142 | |
XM_017024370.1 | XP_016879859.1 | 22 - 23 | 2178 | 142 | |
XM_017024371.1 | XP_016879860.1 | 21 - 22 | 2231 | 142 | |
GenBank mRNA | AB028962.1 | - | 11 - 12 | 1254 | 142 |
AJ628446.1 | - | 22 - 23 | 2200 | 142 | |
AJ628447.1 | - | 21 - 22 | 2155 | 142 | |
AK124640.1 | - | 22 - 23 | 2289 | 142 | |
BC140870.1 | - | 22 - 23 | 2156 | 142 | |
BC140871.1 | - | 22 - 23 | 2156 | 142 | |
CR627427.1 | - | 21 - 22 | 2124 | 142 | |
DN831680.1 | - | 5 - 6 | 509 | 142 |