Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303496.1 | NP_001290425.1 | 3 - 4 | 911 | 105 |
NM_001303497.1 | NP_001290426.1 | 4 - 5 | 740 | 105 | |
NM_001303498.1 | NP_001290427.1 | 3 - 4 | 476 | 105 | |
NM_001303500.1 | NP_001290429.1 | 2 - 3 | 344 | 105 | |
NM_152703.3 | NP_689916.2 | 3 - 4 | 1116 | 105 | |
XM_005250193.3 | XP_005250250.1 | 4 - 5 | 548 | 105 | |
XM_011515903.2 | XP_011514205.1 | 4 - 5 | 801 | 105 | |
XM_017011822.1 | XP_016867311.1 | 2 - 3 | 865 | 105 | |
XM_017011823.1 | XP_016867312.1 | 3 - 4 | 621 | 105 | |
GenBank mRNA | AB095926.1 | - | 3 - 4 | 1084 | 105 |
AF474973.1 | - | 4 - 5 | 493 | 105 | |
AY195582.1 | - | 2 - 3 | 301 | 105 | |
AY195584.1 | - | 4 - 5 | 697 | 105 | |
AY195585.1 | - | 3 - 4 | 868 | 105 | |
AY195586.1 | - | 3 - 4 | 1073 | 105 | |
BC127117.1 | - | 3 - 4 | 352 | 105 | |
BC127118.1 | - | 2 - 3 | 728 | 105 | |
DQ068177.1 | - | 3 - 4 | 433 | 105 |