Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021930.4 | NP_068749.3 | 6 - 7 | 1074 | 95 |
XM_017012496.1 | XP_016867985.1 | 6 - 7 | 853 | 95 | |
XM_017012497.1 | XP_016867986.1 | 6 - 7 | 949 | 95 | |
GenBank mRNA | AF317622.1 | - | 6 - 7 | 953 | 95 |
AK055508.1 | - | 6 - 7 | 989 | 95 | |
AK057757.1 | - | 3 - 4 | 475 | 95 | |
BC068483.1 | - | 6 - 7 | 1075 | 95 |