Assay Design



Gene Transcripts

Gene Symbol
AAMDC
Entrez Gene ID
28971
Gene Name
adipogenesis associated Mth938 domain containing
Gene Aliases
C11orf67, CK067, PTD015
Chromosome Location
Chr.11: 77821114 - 77901281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316957.1 NP_001303886.1 4 - 5 417 69
NM_001316958.1 NP_001303887.1 4 - 5 417 69
XM_017017620.1 XP_016873109.1 3 - 4 397 69
GenBank mRNA AF092136.1 - 4 - 5 376 69
CN362204.1 - 4 - 5 388 69

View Full Product Details