Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022659.3 | NP_073150.2 | 2 - 3 | 559 | 62 |
GenBank mRNA | AK001144.1 | - | 2 - 3 | 612 | 62 |
AL701879.1 | - | 2 - 3 | 208 | 62 | |
BC137106.1 | - | 2 - 3 | 559 | 62 | |
BC137107.1 | - | 2 - 3 | 559 | 62 |