Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322793.1 | NP_001309722.1 | 9 - 10 | 1322 | 80 |
NM_001322794.1 | NP_001309723.1 | 9 - 10 | 1205 | 80 | |
NM_001322795.1 | NP_001309724.1 | 10 - 11 | 1504 | 80 | |
NM_001322796.1 | NP_001309725.1 | 9 - 10 | 1486 | 80 | |
NM_004969.3 | NP_004960.2 | 9 - 10 | 1322 | 80 | |
NR_136399.1 | 9 - 10 | 1322 | 80 | ||
XM_017016187.1 | XP_016871676.1 | 9 - 10 | 1245 | 80 | |
XM_017016188.1 | XP_016871677.1 | 9 - 10 | 1839 | 80 | |
XM_017016189.1 | XP_016871678.1 | 10 - 11 | 1238 | 80 | |
XM_017016190.1 | XP_016871679.1 | 9 - 10 | 1365 | 80 | |
GenBank mRNA | AK312810.1 | - | 9 - 10 | 1247 | 80 |
BC096336.1 | - | 9 - 10 | 1286 | 80 | |
BC096337.1 | - | 9 - 10 | 1286 | 80 | |
BC096339.3 | - | 9 - 10 | 1286 | 80 | |
BX648462.1 | - | 9 - 10 | 1487 | 80 | |
M21188.1 | - | 9 - 10 | 1303 | 80 |