Assay Design



Gene Transcripts

Gene Symbol
CDC42SE1
Entrez Gene ID
56882
Gene Name
CDC42 small effector 1
Gene Aliases
SCIP1, SPEC1
Chromosome Location
Chr.1: 151050971 - 151059649 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038707.1 NP_001033796.1 2 - 3 308 72
GenBank mRNA BX339337.2 - 2 - 3 221 72

View Full Product Details