Assay Design
Gene Transcripts
- Gene Symbol
- EPYC
- Entrez Gene ID
-
1833
- Gene Name
- epiphycan
- Gene Aliases
-
DSPG3, PGLB, Pg-Lb, SLRR3B
- Chromosome Location
-
Chr.12: 90963679 - 91005026 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_004950.4 |
NP_004941.2 |
1 - 2 |
77 |
79 |
GenBank mRNA |
BC030958.1 |
- |
1 - 2 |
77 |
79 |
View Full Product Details