Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282606.1 | NP_001269535.1 | 6 - 7 | 723 | 120 |
NM_001282610.1 | NP_001269539.1 | 5 - 6 | 544 | 120 | |
NM_016056.3 | NP_057140.2 | 5 - 6 | 582 | 120 | |
GenBank mRNA | AF113127.1 | - | 5 - 6 | 504 | 120 |
AF151877.1 | - | 5 - 6 | 521 | 120 | |
AF182041.1 | - | 5 - 6 | 509 | 120 | |
AF182414.1 | - | 6 - 7 | 610 | 120 | |
AK075069.1 | - | 5 - 6 | 496 | 120 | |
AK225158.1 | - | 5 - 6 | 502 | 120 | |
AL117550.1 | - | 5 - 6 | 961 | 120 | |
BC004401.1 | - | 5 - 6 | 499 | 120 | |
BC020613.1 | - | 5 - 6 | 485 | 120 |