Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198964.1 | NP_945315.1 | 1 - 2 | 276 | 76 |
NM_198966.1 | NP_945317.1 | 1 - 2 | 276 | 76 | |
GenBank mRNA | AK313476.1 | - | 1 - 2 | 293 | 76 |
BG676028.1 | - | 1 - 2 | 272 | 76 | |
BG679319.1 | - | 1 - 2 | 276 | 76 | |
J03802.1 | - | 1 - 2 | 276 | 76 |