Assay Design



Gene Transcripts

Gene Symbol
EBPL
Entrez Gene ID
84650
Gene Name
emopamil binding protein like
Gene Aliases
EBRP
Chromosome Location
Chr.13: 49660676 - 49691487 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278636.1 NP_001265565.1 3 - 4 440 132
GenBank mRNA BC021021.1 - 3 - 4 417 132

View Full Product Details