Assay Design



Gene Transcripts

Gene Symbol
FAM53A
Entrez Gene ID
152877
Gene Name
family with sequence similarity 53 member A
Gene Aliases
DNTNP
Chromosome Location
Chr.4: 1574055 - 1684313 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013622.3 NP_001013644.1 3 - 4 335 82
NM_001174070.2 NP_001167541.1 3 - 4 382 82
NM_001297435.1 NP_001284364.1 3 - 4 735 82
XM_005247948.3 XP_005248005.1 3 - 4 1032 82
XM_011513402.2 XP_011511704.1 3 - 4 1038 82
XM_011513403.1 XP_011511705.1 3 - 4 1032 82
XM_011513404.2 XP_011511706.1 3 - 4 1032 82
XM_011513405.1 XP_011511707.1 3 - 4 1032 82
XM_017007802.1 XP_016863291.1 3 - 4 1032 82
GenBank mRNA AK125566.1 - 3 - 4 330 82
BC070112.1 - 3 - 4 335 82
HY040719.1 - 3 - 4 388 82

View Full Product Details