Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012116.3 | NP_036248.3 | 4 - 5 | 846 | 99 |
XM_005258696.3 | XP_005258753.1 | 4 - 5 | 853 | 99 | |
XM_011526688.2 | XP_011524990.1 | 4 - 5 | 854 | 99 | |
GenBank mRNA | AB028645.1 | - | 4 - 5 | 824 | 99 |
AF117646.1 | - | 4 - 5 | 846 | 99 | |
BC028915.1 | - | 4 - 5 | 804 | 99 |