Assay Design



Gene Transcripts

Gene Symbol
DCUN1D2
Entrez Gene ID
55208
Gene Name
defective in cullin neddylation 1 domain containing 2
Gene Aliases
C13orf17
Chromosome Location
Chr.13: 113455819 - 113492047 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017020644.1 XP_016876133.1 1 - 2 41 70
GenBank mRNA BM836766.1 - 1 - 2 43 70

View Full Product Details