Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031689.2 | NP_001026859.1 | 3 - 4 | 869 | 103 |
NM_001321546.1 | NP_001308475.1 | 3 - 4 | 872 | 103 | |
XM_011518072.2 | XP_011516374.1 | 3 - 4 | 858 | 103 | |
XM_017015319.1 | XP_016870808.1 | 3 - 4 | 858 | 103 | |
GenBank mRNA | AF083395.1 | - | 2 - 3 | 273 | 103 |
AF145020.1 | - | 3 - 4 | 668 | 103 | |
AJ238243.1 | - | 2 - 3 | 301 | 103 | |
AK002143.1 | - | 3 - 4 | 632 | 103 | |
AK022761.1 | - | 3 - 4 | 547 | 103 | |
AK223544.1 | - | 2 - 3 | 304 | 103 | |
AL133608.1 | - | 3 - 4 | 862 | 103 | |
BC032551.1 | - | 3 - 4 | 551 | 103 |