Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031689.2 | NP_001026859.1 | 9 - 10 | 1845 | 75 |
XM_011518072.2 | XP_011516374.1 | 9 - 10 | 1834 | 75 | |
GenBank mRNA | AF083395.1 | - | 8 - 9 | 1249 | 75 |
AF145020.1 | - | 9 - 10 | 1644 | 75 | |
AJ238243.1 | - | 8 - 9 | 1277 | 75 | |
AK001642.1 | - | 9 - 10 | 1548 | 75 | |
AK002143.1 | - | 9 - 10 | 1608 | 75 | |
AK022761.1 | - | 9 - 10 | 1523 | 75 | |
AK223544.1 | - | 8 - 9 | 1280 | 75 | |
AL133608.1 | - | 9 - 10 | 1838 | 75 | |
BC032551.1 | - | 9 - 10 | 1527 | 75 |