Assay Design



Gene Transcripts

Gene Symbol
SLX4
Entrez Gene ID
84464
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
BTBD12, FANCP, MUS312
Chromosome Location
Chr.16: 3581181 - 3611598 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032444.2 NP_115820.2 8 - 9 2550 116
XM_011522715.2 XP_011521017.1 8 - 9 2564 116
XM_017023775.1 XP_016879264.1 5 - 6 1955 116
GenBank mRNA AB075867.1 - 6 - 7 3144 116
BC029952.1 - 7 - 8 1994 116

View Full Product Details