Assay Design



Gene Transcripts

Gene Symbol
SLX4
Entrez Gene ID
84464
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
BTBD12, FANCP, MUS312
Chromosome Location
Chr.16: 3581181 - 3611598 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032444.2 NP_115820.2 11 - 12 2952 122
XM_011522715.2 XP_011521017.1 11 - 12 2966 122
XM_017023775.1 XP_016879264.1 8 - 9 2357 122
GenBank mRNA AB058687.1 - - 326 122
BC036335.1 - 2 - 3 344 122

View Full Product Details