Assay Design



Gene Transcripts

Gene Symbol
SLX4
Entrez Gene ID
84464
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
BTBD12, FANCP, MUS312
Chromosome Location
Chr.16: 3581181 - 3611598 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032444.2 NP_115820.2 4 - 5 1579 69
XM_011522715.2 XP_011521017.1 4 - 5 1593 69
GenBank mRNA BC029952.1 - 3 - 4 1023 69

View Full Product Details