Assay Design



Gene Transcripts

Gene Symbol
MROH1
Entrez Gene ID
727957
Gene Name
maestro heat like repeat family member 1
Gene Aliases
HEATR7A
Chromosome Location
Chr.8: 144148016 - 144261940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032450.2 NP_115826.2 24 - 25 2586 65
XM_011517270.2 XP_011515572.1 24 - 25 2724 65
XM_011517271.2 XP_011515573.1 24 - 25 2723 65
XM_011517273.2 XP_011515575.1 24 - 25 2722 65
XM_011517275.2 XP_011515577.1 24 - 25 2722 65
XM_011517276.2 XP_011515578.1 24 - 25 2750 65
XM_011517277.1 XP_011515579.1 24 - 25 2586 65
XM_011517278.1 XP_011515580.1 23 - 24 2466 65
XM_011517279.2 XP_011515581.1 23 - 24 2630 65
XM_011517281.1 XP_011515583.1 23 - 24 2639 65
XM_011517282.1 XP_011515584.1 22 - 23 2564 65
XM_011517283.2 XP_011515585.1 24 - 25 2545 65
XM_017013806.1 XP_016869295.1 21 - 22 2201 65
GenBank mRNA AL834309.1 - 23 - 24 2470 65

View Full Product Details