Assay Design



Gene Transcripts

Gene Symbol
MROH1
Entrez Gene ID
727957
Gene Name
maestro heat like repeat family member 1
Gene Aliases
HEATR7A
Chromosome Location
Chr.8: 144148016 - 144261940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099280.2 NP_001092750.1 5 - 6 544 101
NM_001099281.2 NP_001092751.1 4 - 5 424 101
NM_001288814.1 NP_001275743.1 4 - 5 431 101
NM_032450.2 NP_115826.2 5 - 6 544 101
XM_011517270.2 XP_011515572.1 5 - 6 682 101
XM_011517271.2 XP_011515573.1 5 - 6 681 101
XM_011517272.2 XP_011515574.1 5 - 6 682 101
XM_011517273.2 XP_011515575.1 5 - 6 680 101
XM_011517274.2 XP_011515576.1 5 - 6 680 101
XM_011517275.2 XP_011515577.1 5 - 6 680 101
XM_011517276.2 XP_011515578.1 5 - 6 708 101
XM_011517277.1 XP_011515579.1 5 - 6 544 101
XM_011517278.1 XP_011515580.1 4 - 5 424 101
XM_011517279.2 XP_011515581.1 4 - 5 588 101
XM_011517281.1 XP_011515583.1 4 - 5 597 101
XM_011517282.1 XP_011515584.1 3 - 4 522 101
XM_011517283.2 XP_011515585.1 5 - 6 503 101
XM_011517285.1 XP_011515587.1 4 - 5 424 101
XM_011517286.2 XP_011515588.1 5 - 6 676 101
XM_017013802.1 XP_016869291.1 5 - 6 680 101
XM_017013803.1 XP_016869292.1 5 - 6 679 101
XM_017013804.1 XP_016869293.1 5 - 6 544 101
XM_017013805.1 XP_016869294.1 4 - 5 424 101
XM_017013806.1 XP_016869295.1 2 - 3 159 101
XM_017013807.1 XP_016869296.1 5 - 6 677 101
GenBank mRNA AB058736.1 - 4 - 5 436 101
AK093563.1 - 5 - 6 543 101
AK295740.1 - 4 - 5 424 101
AL834309.1 - 4 - 5 428 101
BC040742.1 - 2 - 3 171 101
BC057824.1 - 5 - 6 524 101
BC067855.1 - 5 - 6 524 101
BC092486.1 - 4 - 5 404 101

View Full Product Details