Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031282.2 | NP_112572.1 | 6 - 7 | 1273 | 69 |
XM_011510034.1 | XP_011508336.1 | 6 - 7 | 1249 | 69 | |
GenBank mRNA | AF329490.1 | - | 6 - 7 | 1137 | 69 |
AF329492.1 | - | 4 - 5 | 878 | 69 | |
AF343659.1 | - | 6 - 7 | 1273 | 69 | |
AF343660.1 | - | 6 - 7 | 1273 | 69 | |
AF343661.1 | - | 6 - 7 | 1273 | 69 | |
AF397452.1 | - | 6 - 7 | 1137 | 69 | |
BC125173.1 | - | 6 - 7 | 1231 | 69 | |
BC125174.1 | - | 6 - 7 | 1231 | 69 | |
BC143628.1 | - | 5 - 6 | 945 | 69 |