Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014856.2 | NP_055671.2 | 26 - 27 | 4679 | 119 |
XM_005245678.4 | XP_005245735.1 | 26 - 27 | 4836 | 119 | |
XM_005245679.2 | XP_005245736.1 | 26 - 27 | 4686 | 119 | |
XM_011510219.1 | XP_011508521.1 | 24 - 25 | 4085 | 119 | |
XM_011510220.2 | XP_011508522.1 | 25 - 26 | 4499 | 119 | |
XM_011510221.2 | XP_011508523.1 | 24 - 25 | 4207 | 119 | |
XM_017002993.1 | XP_016858482.1 | 23 - 24 | 4061 | 119 | |
GenBank mRNA | AB007945.2 | - | 26 - 27 | 4679 | 119 |
AK226053.1 | - | 21 - 22 | 3334 | 119 |