Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015203.4 | NP_056018.2 | 4 - 5 | 1123 | 129 |
XM_005245033.2 | XP_005245090.1 | 4 - 5 | 1086 | 129 | |
XM_006711240.2 | XP_006711303.1 | 4 - 5 | 1086 | 129 | |
XM_011509344.2 | XP_011507646.1 | 4 - 5 | 1086 | 129 | |
XM_017000764.1 | XP_016856253.1 | 4 - 5 | 461 | 129 | |
XM_017000765.1 | XP_016856254.1 | 4 - 5 | 1086 | 129 | |
GenBank mRNA | AB007929.2 | - | 4 - 5 | 492 | 129 |
AK291703.1 | - | 4 - 5 | 541 | 129 | |
BX641025.1 | - | 5 - 6 | 1018 | 129 |