Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297673.1 | NP_001284602.1 | 6 - 7 | 1390 | 158 |
NM_001297674.1 | NP_001284603.1 | 6 - 7 | 1390 | 158 | |
NM_015203.4 | NP_056018.2 | 7 - 8 | 1468 | 158 | |
XM_005245033.2 | XP_005245090.1 | 7 - 8 | 1431 | 158 | |
XM_006711240.2 | XP_006711303.1 | 7 - 8 | 1431 | 158 | |
XM_011509344.2 | XP_011507646.1 | 7 - 8 | 1431 | 158 | |
XM_017000763.1 | XP_016856252.1 | 6 - 7 | 1353 | 158 | |
XM_017000764.1 | XP_016856253.1 | 7 - 8 | 806 | 158 | |
XM_017000765.1 | XP_016856254.1 | 7 - 8 | 1431 | 158 | |
XM_017000766.1 | XP_016856255.1 | 6 - 7 | 1065 | 158 | |
XM_017000767.1 | XP_016856256.1 | 6 - 7 | 1353 | 158 | |
GenBank mRNA | AB007929.2 | - | 7 - 8 | 837 | 158 |
AK056707.1 | - | 7 - 8 | 875 | 158 | |
AK125864.1 | - | 7 - 8 | 1038 | 158 | |
AK291703.1 | - | 7 - 8 | 886 | 158 | |
AK304380.1 | - | 6 - 7 | 808 | 158 | |
AK308148.1 | - | 6 - 7 | 799 | 158 | |
BC045623.1 | - | 6 - 7 | 791 | 158 | |
BX641025.1 | - | 8 - 9 | 1363 | 158 |