Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001493.2 | NP_001484.1 | 2 - 3 | 494 | 62 |
GenBank mRNA | AB102647.1 | - | 2 - 3 | 152 | 62 |
AF400433.1 | - | 2 - 3 | 159 | 62 | |
AK090746.1 | - | 2 - 3 | 242 | 62 | |
AK131466.1 | - | 1 - 2 | 624 | 62 | |
AK294893.1 | - | 2 - 3 | 253 | 62 | |
AK295306.1 | - | 2 - 3 | 253 | 62 | |
AK303248.1 | - | 2 - 3 | 247 | 62 | |
AL833629.1 | - | 2 - 3 | 248 | 62 | |
BC000317.2 | - | 2 - 3 | 194 | 62 | |
BC012201.1 | - | 2 - 3 | 241 | 62 | |
BT019884.1 | - | 2 - 3 | 152 | 62 | |
CR542258.1 | - | 2 - 3 | 152 | 62 | |
CR542276.1 | - | 2 - 3 | 152 | 62 | |
D45021.1 | - | 2 - 3 | 182 | 62 | |
KU178091.1 | - | 2 - 3 | 152 | 62 | |
KU178092.1 | - | 2 - 3 | 152 | 62 | |
X79353.1 | - | 2 - 3 | 232 | 62 |