Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001493.2 | NP_001484.1 | 4 - 5 | 725 | 66 |
GenBank mRNA | AB102647.1 | - | 4 - 5 | 383 | 66 |
AF400433.1 | - | 4 - 5 | 390 | 66 | |
AK090746.1 | - | 4 - 5 | 473 | 66 | |
AK131466.1 | - | 3 - 4 | 855 | 66 | |
AK294893.1 | - | 4 - 5 | 484 | 66 | |
AL833629.1 | - | 4 - 5 | 479 | 66 | |
BC000317.2 | - | 4 - 5 | 425 | 66 | |
BC012201.1 | - | 4 - 5 | 472 | 66 | |
BT019884.1 | - | 4 - 5 | 383 | 66 | |
CR542258.1 | - | 4 - 5 | 383 | 66 | |
CR542276.1 | - | 4 - 5 | 383 | 66 | |
D45021.1 | - | 4 - 5 | 413 | 66 | |
KU178091.1 | - | 4 - 5 | 383 | 66 | |
X79353.1 | - | 4 - 5 | 463 | 66 |