Assay Design



Gene Transcripts

Gene Symbol
GDI1
Entrez Gene ID
2664
Gene Name
GDP dissociation inhibitor 1
Gene Aliases
1A, GDIL, MRX41, MRX48, OPHN2, RABGD1A, RABGDIA, XAP-4
Chromosome Location
Chr.X: 154436913 - 154443467 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001493.2 NP_001484.1 9 - 10 1476 64
GenBank mRNA AB102647.1 - 9 - 10 1134 64
AF400433.1 - 9 - 10 1141 64
AK090746.1 - 9 - 10 1224 64
AK123405.1 - 2 - 3 1414 64
AK303248.1 - 9 - 10 1048 64
AL833629.1 - 9 - 10 1230 64
BC000317.2 - 9 - 10 1176 64
BC012201.1 - 9 - 10 1223 64
BT019884.1 - 9 - 10 1134 64
CR542258.1 - 9 - 10 1134 64
CR542276.1 - 9 - 10 1134 64
D45021.1 - 9 - 10 1164 64
KU178091.1 - 9 - 10 1134 64
KU178092.1 - 7 - 8 766 64
L05086.1 - 1 - 2 155 64
X74608.1 - 1 - 2 86 64
X79353.1 - 9 - 10 1214 64
X87194.1 - 1 - 2 86 64

View Full Product Details