Assay Design



Gene Transcripts

Gene Symbol
DTNA
Entrez Gene ID
1837
Gene Name
dystrobrevin alpha
Gene Aliases
D18S892E, DRP3, DTN, DTN-A, LVNC1
Chromosome Location
Chr.18: 34493290 - 34891844 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198938.1 NP_001185867.1 11 - 12 1430 70
NM_001198939.1 NP_001185868.1 11 - 12 1430 70
NM_001198940.1 NP_001185869.1 11 - 12 1430 70
NM_001198941.1 NP_001185870.1 10 - 11 1298 70
NM_001198943.1 NP_001185872.1 3 - 4 576 70
NM_032975.3 NP_116757.2 12 - 13 1443 70
NM_032979.4 NP_116761.2 12 - 13 1443 70
NM_032981.4 NP_116763.1 3 - 4 517 70
XM_011525853.2 XP_011524155.1 11 - 12 1429 70
XM_017025576.1 XP_016881065.1 12 - 13 1432 70
XM_017025577.1 XP_016881066.1 10 - 11 1298 70
XM_017025584.1 XP_016881073.1 12 - 13 1432 70
XM_017025585.1 XP_016881074.1 12 - 13 1432 70
XM_017025586.1 XP_016881075.1 12 - 13 1432 70
XM_017025587.1 XP_016881076.1 11 - 12 1423 70
XM_017025588.1 XP_016881077.1 10 - 11 1298 70
XM_017025589.1 XP_016881078.1 11 - 12 1429 70
XM_017025590.1 XP_016881079.1 11 - 12 1423 70
XM_017025591.1 XP_016881080.1 10 - 11 1298 70
XM_017025592.1 XP_016881081.1 11 - 12 1429 70
XM_017025593.1 XP_016881082.1 11 - 12 1423 70
XM_017025594.1 XP_016881083.1 10 - 11 1298 70
XM_017025596.1 XP_016881085.1 11 - 12 1429 70
XM_017025597.1 XP_016881086.1 11 - 12 1423 70
XM_017025598.1 XP_016881087.1 10 - 11 1298 70
XM_017025599.1 XP_016881088.1 3 - 4 491 70
GenBank mRNA AB209102.1 - 11 - 12 1383 70
AK291156.1 - 10 - 11 1233 70
AK296450.1 - 3 - 4 577 70
AK308410.1 - 11 - 12 1410 70
AK315215.1 - 3 - 4 408 70
AL833285.1 - 11 - 12 1390 70
BG705393.1 - 3 - 4 500 70
BI668892.1 - 3 - 4 494 70
U26743.1 - 3 - 4 464 70
U26744.1 - 11 - 12 1150 70
U46744.1 - 11 - 12 1156 70

View Full Product Details