Assay Design



Gene Transcripts

Gene Symbol
DTNA
Entrez Gene ID
1837
Gene Name
dystrobrevin alpha
Gene Aliases
D18S892E, DRP3, DTN, DTN-A, LVNC1
Chromosome Location
Chr.18: 34493290 - 34891844 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128175.1 NP_001121647.1 4 - 5 496 78
NM_001198938.1 NP_001185867.1 4 - 5 496 78
NM_001198939.1 NP_001185868.1 4 - 5 496 78
NM_001198940.1 NP_001185869.1 4 - 5 496 78
NM_001198941.1 NP_001185870.1 3 - 4 364 78
NM_001198945.1 NP_001185874.1 4 - 5 500 78
NM_001390.4 NP_001381.2 2 - 3 150 78
NM_001391.5 NP_001382.2 2 - 3 150 78
NM_001392.4 NP_001383.2 4 - 5 500 78
NM_032975.3 NP_116757.2 4 - 5 500 78
NM_032978.6 NP_116760.2 3 - 4 364 78
NM_032979.4 NP_116761.2 4 - 5 500 78
XM_005258221.4 XP_005258278.1 4 - 5 495 78
XM_011525853.2 XP_011524155.1 4 - 5 495 78
XM_017025575.1 XP_016881064.1 3 - 4 364 78
XM_017025576.1 XP_016881065.1 4 - 5 489 78
XM_017025577.1 XP_016881066.1 3 - 4 364 78
XM_017025578.1 XP_016881067.1 4 - 5 489 78
XM_017025579.1 XP_016881068.1 3 - 4 364 78
XM_017025580.1 XP_016881069.1 4 - 5 495 78
XM_017025581.1 XP_016881070.1 4 - 5 489 78
XM_017025582.1 XP_016881071.1 3 - 4 364 78
XM_017025583.1 XP_016881072.1 4 - 5 495 78
XM_017025584.1 XP_016881073.1 4 - 5 489 78
XM_017025585.1 XP_016881074.1 4 - 5 489 78
XM_017025586.1 XP_016881075.1 4 - 5 489 78
XM_017025587.1 XP_016881076.1 4 - 5 489 78
XM_017025588.1 XP_016881077.1 3 - 4 364 78
XM_017025589.1 XP_016881078.1 4 - 5 495 78
XM_017025590.1 XP_016881079.1 4 - 5 489 78
XM_017025591.1 XP_016881080.1 3 - 4 364 78
XM_017025592.1 XP_016881081.1 4 - 5 495 78
XM_017025593.1 XP_016881082.1 4 - 5 489 78
XM_017025594.1 XP_016881083.1 3 - 4 364 78
XM_017025595.1 XP_016881084.1 4 - 5 495 78
XM_017025596.1 XP_016881085.1 4 - 5 495 78
XM_017025597.1 XP_016881086.1 4 - 5 489 78
XM_017025598.1 XP_016881087.1 3 - 4 364 78
XM_017025600.1 XP_016881089.1 3 - 4 364 78
XM_017025601.1 XP_016881090.1 4 - 5 489 78
XM_017025602.1 XP_016881091.1 3 - 4 364 78
GenBank mRNA AB209102.1 - 4 - 5 449 78
AK291156.1 - 3 - 4 299 78
AK294746.1 - 4 - 5 486 78
AK308410.1 - 4 - 5 475 78
AL833285.1 - 4 - 5 456 78
BC005300.1 - 4 - 5 456 78
BG250424.1 - - 407 78
BT006937.1 - 2 - 3 149 78
DA073612.1 - 4 - 5 500 78
DA197970.1 - 4 - 5 496 77
DA398530.1 - 3 - 4 364 78
U26742.1 - 4 - 5 473 78
U26744.1 - 3 - 4 207 78
U46744.1 - 3 - 4 213 78
U46745.1 - 3 - 4 420 78

View Full Product Details