Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171930.1 | NP_001165401.1 | 10 - 11 | 1335 | 59 |
NM_001171931.1 | NP_001165402.1 | 10 - 11 | 1335 | 59 | |
NM_001171932.1 | NP_001165403.1 | 10 - 11 | 1335 | 59 | |
NM_022124.5 | NP_071407.4 | 10 - 11 | 1335 | 59 | |
NM_052836.3 | NP_443068.1 | 10 - 11 | 1335 | 59 | |
XM_006717940.3 | XP_006718003.1 | 10 - 11 | 1329 | 59 | |
XM_006717942.3 | XP_006718005.1 | 10 - 11 | 1329 | 59 | |
XM_011540039.2 | XP_011538341.1 | 10 - 11 | 1329 | 59 | |
XM_011540042.2 | XP_011538344.1 | 10 - 11 | 1329 | 59 | |
XM_011540043.2 | XP_011538345.1 | 10 - 11 | 1329 | 59 | |
XM_011540044.2 | XP_011538346.1 | 10 - 11 | 970 | 59 | |
XM_011540045.2 | XP_011538347.1 | 10 - 11 | 1329 | 59 | |
XM_011540046.2 | XP_011538348.1 | 5 - 6 | 764 | 59 | |
XM_011540048.2 | XP_011538350.1 | 10 - 11 | 1329 | 59 | |
XM_011540049.2 | XP_011538351.1 | 10 - 11 | 1329 | 59 | |
XM_011540051.2 | XP_011538353.1 | 10 - 11 | 1329 | 59 | |
XM_017016499.1 | XP_016871988.1 | 10 - 11 | 1335 | 59 | |
XM_017016500.1 | XP_016871989.1 | 10 - 11 | 1329 | 59 | |
GenBank mRNA | AF312024.1 | - | 9 - 10 | 945 | 59 |
AK074160.1 | - | 3 - 4 | 317 | 59 | |
AY010111.2 | - | 9 - 10 | 945 | 59 | |
AY358617.1 | - | 10 - 11 | 985 | 59 | |
BC011570.1 | - | 10 - 11 | 1340 | 59 | |
BC032581.1 | - | 10 - 11 | 1341 | 59 | |
BC065284.1 | - | 10 - 11 | 1312 | 59 | |
BC108254.1 | - | 8 - 9 | 1103 | 59 | |
BC136976.1 | - | 10 - 11 | 1205 | 59 | |
BC136977.1 | - | 10 - 11 | 1204 | 59 | |
BC139903.1 | - | 10 - 11 | 1327 | 59 |