Assay Design



Gene Transcripts

Gene Symbol
CDH23
Entrez Gene ID
64072
Gene Name
cadherin-related 23
Gene Aliases
CDHR23, USH1D
Chromosome Location
Chr.10: 71396934 - 71815947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171933.1 NP_001165404.1 17 - 18 2877 97
NM_001171934.1 NP_001165405.1 17 - 18 2877 97
NM_022124.5 NP_071407.4 64 - 65 9670 97
XM_006717940.3 XP_006718003.1 65 - 66 9724 97
XM_006717942.3 XP_006718005.1 63 - 64 9658 97
XM_011540039.2 XP_011538341.1 64 - 65 9721 97
XM_011540042.2 XP_011538344.1 66 - 67 9634 97
XM_011540043.2 XP_011538345.1 65 - 66 9724 97
XM_011540044.2 XP_011538346.1 65 - 66 9365 97
XM_011540045.2 XP_011538347.1 65 - 66 9724 97
XM_011540046.2 XP_011538348.1 60 - 61 9159 97
XM_011540047.2 XP_011538349.1 54 - 55 8459 97
XM_011540052.2 XP_011538354.1 37 - 38 5999 97
XM_017016499.1 XP_016871988.1 64 - 65 9670 97
XM_017016500.1 XP_016871989.1 64 - 65 9598 97
XM_017016501.1 XP_016871990.1 44 - 45 7226 97
XM_017016502.1 XP_016871991.1 44 - 45 7414 97
XM_017016503.1 XP_016871992.1 34 - 35 5402 97
XM_017016504.1 XP_016871993.1 28 - 29 5187 97
XM_017016505.1 XP_016871994.1 28 - 29 5218 97
XM_017016506.1 XP_016871995.1 27 - 28 4657 97
XM_017016507.1 XP_016871996.1 21 - 22 3479 97
XM_017016508.1 XP_016871997.1 17 - 18 2932 97
GenBank mRNA AB053445.1 - 16 - 17 2816 97
AF312024.1 - 63 - 64 9280 97
AY010111.2 - 63 - 64 9280 97
AY563165.1 - 17 - 18 2877 97
AY563166.1 - 17 - 18 2877 97

View Full Product Details