Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002428.3 | NP_002419.1 | 4 - 5 | 1535 | 64 |
GenBank mRNA | BC006412.1 | - | 4 - 5 | 655 | 64 |
BC036495.1 | - | 4 - 5 | 1541 | 64 | |
BC055428.1 | - | 4 - 5 | 1536 | 64 | |
BT006715.1 | - | 2 - 3 | 435 | 64 | |
D85510.1 | - | 1 - 2 | 265 | 64 | |
D86331.1 | - | 3 - 4 | 473 | 64 | |
Z48482.1 | - | 4 - 5 | 798 | 64 |