Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000876.3 | NP_000867.2 | 17 - 18 | 2490 | 102 |
GenBank mRNA | AB209668.1 | - | 16 - 17 | 2113 | 102 |
J03528.1 | - | 17 - 18 | 2489 | 102 | |
Y00285.1 | - | 17 - 18 | 2489 | 102 |