Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000876.3 | NP_000867.2 | 24 - 25 | 3555 | 70 |
GenBank mRNA | AB209668.1 | - | 23 - 24 | 3178 | 70 |
J03528.1 | - | 24 - 25 | 3554 | 70 | |
Y00285.1 | - | 24 - 25 | 3554 | 70 |