Assay Design



Gene Transcripts

Gene Symbol
IGF2R
Entrez Gene ID
3482
Gene Name
insulin like growth factor 2 receptor
Gene Aliases
CD222, CI-M6PR, CIMPR, M6P-R, M6P/IGF2R, MPR 300, MPR1, MPR300, MPRI
Chromosome Location
Chr.6: 159969099 - 160111504 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000876.3 NP_000867.2 3 - 4 561 72
GenBank mRNA AB209668.1 - 2 - 3 184 72
J03528.1 - 3 - 4 560 72
Y00285.1 - 3 - 4 560 72

View Full Product Details