Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166247.1 | NP_001159719.1 | 1 - 2 | 412 | 94 |
NM_021956.4 | NP_068775.1 | 1 - 2 | 412 | 94 | |
NM_175768.3 | NP_786944.1 | 1 - 2 | 412 | 94 | |
XM_005266945.2 | XP_005267002.1 | 2 - 3 | 542 | 94 | |
XM_011535777.2 | XP_011534079.1 | 1 - 2 | 579 | 94 | |
XM_011535778.2 | XP_011534080.1 | 1 - 2 | 579 | 94 | |
XM_017010781.1 | XP_016866270.1 | 1 - 2 | 579 | 94 | |
XM_017010782.1 | XP_016866271.1 | 1 - 2 | 579 | 94 | |
GenBank mRNA | AJ301610.1 | - | 1 - 2 | 119 | 94 |
AK289542.1 | - | 1 - 2 | 346 | 94 | |
BC037954.1 | - | 1 - 2 | 604 | 94 | |
BC063814.1 | - | 1 - 2 | 322 | 94 | |
BQ717946.1 | - | 1 - 2 | 319 | 94 | |
HM149335.1 | - | 1 - 2 | 119 | 94 | |
U16126.1 | - | 1 - 2 | 119 | 94 |