Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166247.1 | NP_001159719.1 | 11 - 12 | 2034 | 81 |
NM_021956.4 | NP_068775.1 | 11 - 12 | 2034 | 81 | |
NM_175768.3 | NP_786944.1 | 11 - 12 | 2034 | 81 | |
XM_005266945.2 | XP_005267002.1 | 12 - 13 | 2164 | 81 | |
XM_005266946.3 | XP_005267003.1 | 11 - 12 | 2119 | 81 | |
XM_011535777.2 | XP_011534079.1 | 11 - 12 | 2201 | 81 | |
XM_011535778.2 | XP_011534080.1 | 11 - 12 | 2201 | 81 | |
XM_017010781.1 | XP_016866270.1 | 11 - 12 | 2201 | 81 | |
GenBank mRNA | AJ252246.1 | - | 11 - 12 | 1741 | 81 |
AJ301610.1 | - | 11 - 12 | 1741 | 81 | |
AK303721.1 | - | 11 - 12 | 1744 | 81 | |
HM149335.1 | - | 11 - 12 | 1741 | 81 | |
HM149336.1 | - | 11 - 12 | 1741 | 81 | |
HM149337.1 | - | 11 - 12 | 1741 | 81 | |
U16126.1 | - | 11 - 12 | 1741 | 81 |