Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016224.4 | NP_057308.1 | 10 - 11 | 1344 | 66 |
XM_005267015.2 | XP_005267072.1 | 12 - 13 | 1422 | 66 | |
XM_011535886.2 | XP_011534188.1 | 7 - 8 | 883 | 66 | |
GenBank mRNA | AB209736.1 | - | 6 - 7 | 675 | 66 |
AF076957.1 | - | 5 - 6 | 564 | 66 | |
AF121859.1 | - | 10 - 11 | 1104 | 66 | |
AF131214.1 | - | 10 - 11 | 1123 | 66 | |
AF172847.1 | - | 10 - 11 | 1224 | 66 | |
AK026637.1 | - | 7 - 8 | 889 | 66 | |
AK127345.1 | - | 7 - 8 | 1199 | 66 | |
AK314360.1 | - | 10 - 11 | 1178 | 66 | |
BC001084.2 | - | 1 - 2 | 145 | 66 | |
BC005022.1 | - | 10 - 11 | 1251 | 66 |