Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016224.4 | NP_057308.1 | 15 - 16 | 1796 | 121 |
XM_005267015.2 | XP_005267072.1 | 17 - 18 | 1874 | 121 | |
XM_011535886.2 | XP_011534188.1 | 12 - 13 | 1335 | 121 | |
GenBank mRNA | AF076957.1 | - | 10 - 11 | 1016 | 121 |
AF121859.1 | - | 15 - 16 | 1556 | 121 | |
AF131214.1 | - | 15 - 16 | 1575 | 121 | |
AF172847.1 | - | 15 - 16 | 1676 | 121 | |
AK026637.1 | - | 12 - 13 | 1341 | 121 | |
AK127345.1 | - | 12 - 13 | 1651 | 121 | |
AK314360.1 | - | 15 - 16 | 1630 | 121 | |
BC001084.2 | - | 6 - 7 | 597 | 121 | |
BC005022.1 | - | 15 - 16 | 1703 | 121 |