Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100398.1 | NP_001093868.1 | 9 - 10 | 775 | 98 |
NM_015085.4 | NP_055900.4 | 10 - 11 | 820 | 98 | |
XM_005256542.2 | XP_005256599.1 | 9 - 10 | 1212 | 98 | |
XM_006721477.3 | XP_006721540.1 | 10 - 11 | 761 | 98 | |
XM_011523738.1 | XP_011522040.1 | 11 - 12 | 873 | 98 | |
XM_011523739.1 | XP_011522041.1 | 10 - 11 | 828 | 98 | |
XM_011523740.2 | XP_011522042.1 | 10 - 11 | 733 | 98 | |
XM_011523741.2 | XP_011522043.1 | 10 - 11 | 909 | 98 | |
XM_011523742.2 | XP_011522044.1 | 7 - 8 | 615 | 98 | |
XM_011523743.2 | XP_011522045.1 | 8 - 9 | 693 | 98 | |
XM_011523744.2 | XP_011522046.1 | 3 - 4 | 281 | 98 | |
XM_011523745.2 | XP_011522047.1 | 3 - 4 | 238 | 98 | |
XM_017024370.1 | XP_016879859.1 | 10 - 11 | 798 | 98 | |
XM_017024371.1 | XP_016879860.1 | 9 - 10 | 851 | 98 | |
GenBank mRNA | AJ628446.1 | - | 10 - 11 | 820 | 98 |
AJ628447.1 | - | 9 - 10 | 775 | 98 | |
AK124640.1 | - | 10 - 11 | 909 | 98 | |
BC140870.1 | - | 10 - 11 | 776 | 98 | |
BC140871.1 | - | 10 - 11 | 776 | 98 | |
CR627427.1 | - | 9 - 10 | 744 | 98 |