Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128833.1 | NP_001122305.1 | 2 - 3 | 241 | 154 |
NM_020899.3 | NP_065950.2 | 2 - 3 | 317 | 154 | |
XM_006721563.3 | XP_006721626.1 | 2 - 3 | 168 | 154 | |
XM_006721564.2 | XP_006721627.1 | 2 - 3 | 148 | 154 | |
XM_011523972.2 | XP_011522274.1 | 2 - 3 | 600 | 154 | |
GenBank mRNA | AB040971.2 | - | 2 - 3 | 241 | 154 |
AK122971.1 | - | 2 - 3 | 141 | 154 | |
AY302699.1 | - | 2 - 3 | 157 | 154 | |
BC043352.2 | - | 2 - 3 | 266 | 154 |